siRNA Details
Gene Symbol: Map4k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106329.1 | NP_001099799.1 | 24 | 1879 | ||
XM_006230753.2 | XP_006230815.1 | 25 | 1850 | |||
XM_006230754.2 | XP_006230816.1 | 25 | 1823 | |||
XM_008760109.1 | XP_008758331.1 | 24 | 1703 | |||
GenBank mRNA | FQ211076.1 | - | 25 | 1820 |
