siRNA Details
Gene Symbol: Ubxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034829.1 | NP_001030001.1 | 4 | 253 | ||
XM_006230975.1 | XP_006231037.1 | 5 | 430 | |||
GenBank mRNA | AJ277751.1 | - | 4 | 253 |
