siRNA Details
Gene Symbol: Eef1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004223.2 | NP_001004223.1 | 2 | 207 | ||
GenBank mRNA | BC079398.1 | - | 3 | 216 | 7112627 | |
BC098895.1 | - | 2 | 207 | 7367246 | ||
FQ212596.1 | - | 3 | 225 | |||
FQ213179.1 | - | 3 | 223 | |||
FQ214210.1 | - | 3 | 223 | |||
FQ214443.1 | - | 3 | 224 | |||
FQ215853.1 | - | 3 | 253 | |||
FQ218274.1 | - | 3 | 223 | |||
FQ219298.1 | - | 3 | 225 | |||
FQ222930.1 | - | 3 | 223 | |||
FQ223614.1 | - | 3 | 223 | |||
FQ224956.1 | - | 2 | 182 | |||
FQ228703.1 | - | 3 | 223 | |||
FQ228918.1 | - | 3 | 224 | |||
FQ230995.1 | - | 3 | 223 | |||
FQ231923.1 | - | 3 | 223 | |||
FQ233868.1 | - | 3 | 223 | |||
FQ234035.1 | - | 3 | 223 | |||
FQ235034.1 | - | 3 | 373 |
Gene Symbol: Slamf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250250.2 | XP_006250312.1 | 7 | 2499 | ||
GenBank mRNA | FQ230857.1 | - | 3 | 1433 |
