siRNA Details
Gene Symbol: Incenp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106335.1 | NP_001099805.1 | 17 | 2570 | ||
XM_006231024.1 | XP_006231086.1 | 18 | 2541 | |||
XM_006231025.1 | XP_006231087.1 | 17 | 2529 | |||
XM_006231026.1 | XP_006231088.1 | 18 | 2517 | |||
XM_006231027.2 | XP_006231089.1 | 18 | 3305 | |||
XM_006231028.2 | XP_006231090.1 | 18 | 2540 |
