siRNA Details
Gene Symbol: Incenp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106335.1 | NP_001099805.1 | 2 | 186 | ||
XM_006231024.1 | XP_006231086.1 | 2 | 145 | |||
XM_006231025.1 | XP_006231087.1 | 2 | 145 | |||
XM_006231026.1 | XP_006231088.1 | 2 | 145 | |||
XM_006231027.2 | XP_006231089.1 | 2 | 909 | |||
XM_006231028.2 | XP_006231090.1 | 2 | 144 |
