siRNA Details
Gene Symbol: Best1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011940.1 | NP_001011940.1 | 9, 10 | 1248 | ||
XM_006231029.2 | XP_006231091.1 | 9, 10 | 1288 | |||
GenBank mRNA | BC079048.1 | - | 9, 10 | 1248 | 7115295 |
