siRNA Details
Gene Symbol: Ms4a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231100.2 | XP_006231162.1 | 3 | 1137 | ||
XM_008760272.1 | XP_008758494.1 | 3 | 1030 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
