siRNA Details
Gene Symbol: Ms4a6bl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006975.1 | NP_001006976.1 | 2 | 162 | ||
GenBank mRNA | BC083691.1 | - | 2 | 162 | 7189412 | |
FQ230476.1 | - | 166 |
Gene Symbol: LOC100912573 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231109.2 | XP_006231171.1 | 214 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
