siRNA Details
Gene Symbol: Lpxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009649.1 | NP_001009649.1 | 2 | 140 | ||
GenBank mRNA | BC088217.1 | - | 2 | 140 | 7384057 | |
FQ230386.1 | - | 2 | 105 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
