siRNA Details
Gene Symbol: Bcap31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004224.1 | NP_001004224.1 | 4 | 306 | ||
XM_006229556.2 | XP_006229618.1 | 3 | 263 | |||
XM_006229557.2 | XP_006229619.1 | 4 | 380 | |||
GenBank mRNA | BC079182.1 | - | 4 | 306 | 7130880 | |
FQ219566.1 | - | 4 | 297 | |||
FQ219790.1 | - | 4 | 305 | |||
FQ230132.1 | - | 4 | 297 |
