siRNA Details
Gene Symbol: Cdc37l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011941.1 | NP_001011941.1 | 4 | 656 | ||
XM_006231201.2 | XP_006231263.1 | 5 | 815 | |||
XM_006231202.2 | XP_006231264.1 | 5 | 815 | |||
XM_008760322.1 | XP_008758544.1 | 5 | 815 | |||
GenBank mRNA | BC083761.1 | - | 4 | 656 | 7192975 | |
FQ232043.1 | - | 3 | 428 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
