siRNA Details
Gene Symbol: Plgrkt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106347.1 | NP_001099817.1 | 4, 5 | 514 | ||
XM_006231203.2 | XP_006231265.1 | 5, 6 | 575 | |||
XM_006231204.2 | XP_006231266.1 | 5, 6 | 479 | |||
XM_006231205.2 | XP_006231267.1 | 4, 5 | 418 | |||
XM_008760323.1 | XP_008758545.1 | 2, 3 | 604 |
