siRNA Details
Gene Symbol: Bloc1s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037349.2 | NP_001032426.1 | 4 | 395 | ||
XM_008760400.1 | XP_008758622.1 | 4 | 422 | |||
XM_008760401.1 | XP_008758623.1 | 5 | 497 | |||
GenBank mRNA | AY786315.1 | - | 4 | 376 | ||
EF061226.2 | - | 5 | 451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
