siRNA Details
Gene Symbol: Arhgap18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170575.1 | NP_001164046.1 | 7 | 1107 | ||
XM_006227680.2 | XP_006227742.1 | 7 | 1893 | |||
GenBank mRNA | CV116120.1 | - | 5 | 590 | ||
FM089382.1 | - | 1 | 53 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
