siRNA Details
Gene Symbol: Arhgap18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170575.1 | NP_001164046.1 | 15 | 1997 | ||
XM_006227680.2 | XP_006227742.1 | 15 | 2783 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
