siRNA Details
Gene Symbol: Morn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024975.1 | NP_001020146.1 | 4 | 397 | ||
XM_008760402.1 | XP_008758624.1 | 6 | 578 | |||
GenBank mRNA | BC091345.1 | - | 4 | 397 | 7316315 | |
EF688599.1 | - | 4 | 397 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_198108, BC057060, AK154147, CV563048, AK159230.
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
