siRNA Details
Gene Symbol: Zfand5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106356.1 | NP_001099826.1 | 1 | 591 | ||
XM_008760297.1 | XP_008758519.1 | 3 | 790 | |||
GenBank mRNA | BC168882.1 | - | 3 | 258 | 9026652 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
