siRNA Details
Gene Symbol: Cyp2c6v1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013904.1 | NP_001013926.1 | 1 | 136 | ||
XM_008759612.1 | XP_008757834.1 | 1 | 63 | |||
XM_008759613.1 | XP_008757835.1 | 1 | 165 | |||
XM_008759614.1 | XP_008757836.1 | 1 | 165 | |||
XM_008759615.1 | XP_008757837.1 | 1 | 165 | |||
GenBank mRNA | BC089886.1 | - | 1 | 136 | 7370532 | |
FQ210770.1 | - | 1 | 84 | |||
K03501.1 | - | 111 | ||||
M13711.1 | - | 1 | 111 | |||
M18336.1 | - | 1 | 146 |
