siRNA Details
Gene Symbol: Ndufb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106360.2 | NP_001099830.1 | 4 | 385 | ||
GenBank mRNA | BC166889.1 | - | 4 | 385 | 6891455 | |
BC166905.1 | - | 4 | 385 | 6887475 | ||
BC166906.1 | - | 4 | 385 | 6888064 | ||
FQ217125.1 | - | 4 | 379 | |||
FQ217646.1 | - | 4 | 388 | |||
FQ223847.1 | - | 4 | 400 |
