siRNA Details
Gene Symbol: BEST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004183.3 | NP_004174.1 | 2 | 668 | ||
NR_134580.1 | 2 | 668 | ||||
XM_005274210.3 | XP_005274267.1 | 2 | 778 | |||
XM_005274219.3 | XP_005274276.1 | 2 | 778 | |||
XM_005274221.3 | XP_005274278.1 | 2 | 778 | |||
XM_011545229.2 | XP_011543531.1 | 2 | 218 | |||
XM_017018229.1 | XP_016873718.1 | 2 | 778 | |||
GenBank mRNA | AF057169.1 | - | 2 | 192 | ||
AF057170.1 | - | 2 | 192 | |||
AF073501.1 | - | 2 | 176 | |||
AK289681.1 | - | 2 | 196 | |||
AK294577.1 | - | 2 | 196 | |||
AK295580.1 | - | 2 | 201 | |||
AY515704.1 | - | 1 | 88 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
