siRNA Details
Gene Symbol: Pdzd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106362.2 | NP_001099832.2 | 15 | 2814 | ||
XM_006231449.2 | XP_006231511.1 | 16 | 3420 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001195265, BC147399, BC096488, BC172054, AK033653, and others.
