siRNA Details
Gene Symbol: Cuedc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079886.1 | NP_001073355.1 | 9 | 1279 | ||
XM_006231458.2 | XP_006231520.1 | 9 | 869 | |||
XM_006231459.2 | XP_006231521.1 | 9 | 1332 | |||
XM_006231460.2 | XP_006231522.1 | 9 | 875 | |||
XM_006231462.2 | XP_006231524.2 | 9 | 1688 | |||
XM_006231465.2 | XP_006231527.1 | 9 | 815 | |||
XM_006231466.2 | XP_006231528.1 | 9 | 821 | |||
XM_008760408.1 | XP_008758630.1 | 9 | 1685 | |||
XM_008760409.1 | XP_008758631.1 | 9 | 1500 | |||
XM_008760410.1 | XP_008758632.1 | 8 | 1603 | |||
XM_008760411.1 | XP_008758633.1 | 9 | 1725 | |||
XM_008760412.1 | XP_008758634.1 | 10 | 918 | |||
GenBank mRNA | BC127518.1 | - | 9 | 1279 | 7104051 | |
FQ220195.1 | - | 9 | 783 | |||
FQ229647.1 | - | 8 | 728 |
