siRNA Details
Gene Symbol: Calhm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008306.1 | NP_001008307.1 | 2 | 613 | ||
GenBank mRNA | BC086540.1 | - | 2 | 613 | 7122173 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
