siRNA Details
Gene Symbol: Col17a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106366.1 | NP_001099836.1 | 6, 7 | 679 | ||
XM_006231476.2 | XP_006231538.1 | 6, 7 | 1425 |

More Information
Other Species Targeted

Gene Ontology Categories:
