siRNA Details
Gene Symbol: Pik3ap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106368.1 | NP_001099838.1 | 5 | 926 | ||
XM_008760414.1 | XP_008758636.1 | 5 | 1568 | |||
XM_008760416.1 | XP_008758638.1 | 4 | 1458 | |||
GenBank mRNA | BC169055.1 | - | 5 | 953 | 7930052 |
