siRNA Details
Gene Symbol: LOC499339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025036.1 | NP_001020207.1 | 16 | 1728 | ||
XM_006231246.2 | XP_006231308.1 | 16 | 1775 | |||
GenBank mRNA | BC092579.1 | - | 16 | 1728 | 7323921 | |
BC098744.1 | - | 15 | 1702 | 7387202 | ||
FQ234057.1 | - | 6 | 573 |
