siRNA Details
Gene Symbol: Ifit1lb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760355.1 | XP_008758577.1 | 2 | 2287 | ||
XM_220058.5 | XP_220058.2 | 2 | 3183 |

More Information
Other Species Targeted

