siRNA Details
Gene Symbol: Ifit1lb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760355.1 | XP_008758577.1 | 2 | 2278 | ||
XM_220058.5 | XP_220058.2 | 2 | 3174 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
