siRNA Details
Gene Symbol: Lipk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106374.1 | NP_001099844.1 | 3 | 275 | ||
XM_008760325.1 | XP_008758547.1 | 4 | 283 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
