siRNA Details
Gene Symbol: Papss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106375.2 | NP_001099845.2 | 3 | 334 | ||
XM_006231274.2 | XP_006231336.1 | 3 | 479 | |||
GenBank mRNA | BC169061.1 | - | 3 | 334 | 5621354 |
