siRNA Details
Gene Symbol: Moxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_220095.7 | XP_220095.4 | 8 | 1300 | ||
GenBank mRNA | BC091331.1 | - | 6 | 684 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Moxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_220095.7 | XP_220095.4 | 8 | 1300 | ||
GenBank mRNA | BC091331.1 | - | 6 | 684 |