siRNA Details
Gene Symbol: LOC294154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039607.1 | NP_001034696.1 | 4 | 606 | ||
XM_008772799.1 | XP_008771021.1 | 4 | 800 | |||
XM_008772800.1 | XP_008771022.1 | 4 | 914 | |||
XM_008772801.1 | XP_008771023.1 | 4 | 1339 | |||
GenBank mRNA | BC112320.1 | - | 4 | 606 | 7933524 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
