siRNA Details
Gene Symbol: RGD1302996 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213610.3 | NP_998775.2 | 3, 4 | 626 | ||
XM_006255918.2 | XP_006255980.1 | 3, 4 | 644 | |||
XM_008772712.1 | XP_008770934.1 | 3, 4 | 515 | |||
XM_008772713.1 | XP_008770935.1 | 3, 4 | 503 | |||
GenBank mRNA | BC088194.1 | - | 3, 4 | 626 | 7373688 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
