siRNA Details
Gene Symbol: Gtf2h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212501.2 | NP_997666.1 | 2 | 233 | ||
XM_006255920.2 | XP_006255982.1 | 2 | 282 | |||
GenBank mRNA | BC083703.1 | - | 2 | 191 | ||
BC127469.1 | - | 2 | 205 | 7324889 | ||
CK472761.1 | - | 2 | 233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
