siRNA Details
Gene Symbol: Ddah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165936.1 | NP_001159408.1 | 2, 3 | 505 | ||
NM_212532.2 | NP_997697.1 | 2, 3 | 552 | |||
XM_008772714.1 | XP_008770936.1 | 3, 4 | 651 | |||
XM_008772715.1 | XP_008770937.1 | 2, 3 | 814 | |||
GenBank mRNA | BC086443.1 | - | 2, 3 | 453 | 7303579 | |
BC097930.1 | - | 2, 3 | 542 | 7388716 | ||
CO385374.1 | - | 2, 3 | 552 | |||
EV770973.1 | - | 2, 3 | 505 |
