siRNA Details
Gene Symbol: Ncr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181822.2 | NP_861543.2 | 3, 4 | 700 | ||
GenBank mRNA | AY273824.1 | - | 3, 4 | 821 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
