siRNA Details
Gene Symbol: Nelfe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165901.1 | NP_001159373.1 | 8, 9 | 966 | ||
NM_212548.2 | NP_997713.1 | 8, 9 | 998 | |||
XM_006255921.2 | XP_006255983.1 | 7, 8 | 1127 | |||
XM_006255922.2 | XP_006255984.1 | 8, 9 | 1176 | |||
GenBank mRNA | BC079427.1 | - | 8, 9 | 998 | 7114415 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001045864, NM_138580, NM_001045863, DV662511, AK012389, and others.
Gene Ontology Categories:
