siRNA Details
Gene Symbol: Skiv2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213559.2 | NP_998724.1 | 21 | 2564 | ||
XM_006255923.1 | XP_006255985.1 | 21 | 2627 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
