siRNA Details
Gene Symbol: Slc39a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008885.2 | NP_001008885.1 | 2, 3 | 886 | ||
NM_001164744.1 | NP_001158216.1 | 3, 4 | 627 | |||
GenBank mRNA | BC079141.1 | - | 2, 3 | 886 | 7127902 | |
DY547565.1 | - | 3, 4 | 627 | |||
FQ212335.1 | - | 3, 4 | 594 | |||
FQ212833.1 | - | 2, 3 | 914 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
