siRNA Details
Gene Symbol: Rps18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213557.1 | NP_998722.1 | 2 | 91 | ||
GenBank mRNA | BC126072.1 | - | 2 | 65 | 8362450 | |
FQ209845.1 | - | 2 | 89 | |||
FQ210672.1 | - | 2 | 73 | |||
FQ210792.1 | - | 2 | 89 | |||
FQ217732.1 | - | 2 | 89 | |||
FQ220696.1 | - | 2 | 89 | |||
FQ221730.1 | - | 2 | 89 | |||
FQ221918.1 | - | 2 | 89 | |||
FQ222652.1 | - | 2 | 89 | |||
FQ222675.1 | - | 2 | 87 | |||
FQ223127.1 | - | 2 | 89 | |||
FQ223169.1 | - | 2 | 90 | |||
FQ223198.1 | - | 2 | 89 | |||
FQ223436.1 | - | 2 | 89 | |||
FQ223481.1 | - | 2 | 89 | |||
X57529.1 | - | 2 | 83 |
Gene Symbol: LOC100360679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002727833.4 | XP_002727879.1 | 1 | 83 |
