siRNA Details
Gene Symbol: Rgl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212547.3 | NP_997712.1 | 9 | 1298 | ||
XM_008772722.1 | XP_008770944.1 | 9 | 1882 | |||
GenBank mRNA | BC107439.1 | - | 9 | 1269 | 7121126 | |
FQ211603.1 | - | 9 | 1304 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009059, AK144440, U54639, AK215408, AK204851, and others.
Gene Ontology Categories:
