siRNA Details
Gene Symbol: Cuta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164706.1 | NP_001158178.1 | 5, 6 | 449 | ||
NM_001164707.1 | NP_001158179.1 | 4, 5 | 502 | |||
NM_212494.2 | NP_997659.2 | 5, 6 | 502 | |||
XM_006256120.2 | XP_006256182.1 | 6, 7 | 533 | |||
GenBank mRNA | BP501235.1 | - | 4, 5 | 516 | ||
CB586341.1 | - | 5, 6 | 449 | |||
DV717217.1 | - | 4, 5 | 474 | |||
FQ220708.1 | - | 5, 6 | 736 | |||
FQ223106.1 | - | 3, 4 | 176 | |||
FQ229047.1 | - | 3, 4 | 176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
