siRNA Details
Gene Symbol: Cuta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164706.1 | NP_001158178.1 | 3, 4 | 355 | ||
NM_001164707.1 | NP_001158179.1 | 2, 3 | 408 | |||
NM_212494.2 | NP_997659.2 | 3, 4 | 408 | |||
XM_006256120.2 | XP_006256182.1 | 4, 5 | 439 | |||
GenBank mRNA | BP501235.1 | - | 2, 3 | 422 | ||
CB586341.1 | - | 3, 4 | 355 | |||
DV717217.1 | - | 2, 3 | 380 | |||
FQ220708.1 | - | 3, 4 | 642 | |||
FQ223106.1 | - | 1, 2 | 82 | |||
FQ229047.1 | - | 1, 2 | 82 |

More Information
Other Species Targeted

Gene Ontology Categories:
