siRNA Details
Gene Symbol: Btbd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013073.1 | NP_001013091.1 | 6 | 1186 | ||
XM_006256185.2 | XP_006256247.1 | 6 | 1212 | |||
GenBank mRNA | BC087068.1 | - | 6 | 1186 | 7133235 |
