siRNA Details
Gene Symbol: Ube2g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106380.1 | NP_001099850.1 | 3 | 186 | ||
GenBank mRNA | BC166733.1 | - | 3 | 119 | 7936238 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_019803, AF296657, BC010321, AK136365, AK154464, and others.
Gene Ontology Categories:
