siRNA Details
Gene Symbol: LOC100909869 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256328.2 | XP_006256390.1 | 3 | 524 |
Gene Symbol: LOC100909461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773851.1 | XP_008772073.1 | 1 | 1679 |
Gene Symbol: LOC103690173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773852.1 | XP_008772074.1 | 3 | 524 |
Gene Symbol: Zfp280b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106384.1 | NP_001099854.1 | 2 | 435 | ||
XM_008772876.1 | XP_008771098.1 | 3 | 524 |
