siRNA Details
Gene Symbol: Asf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106389.1 | NP_001099859.1 | 2 | 233 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025541, AK088618, AK007804, AK012376, AK010210, and others.
Gene Ontology Categories:
