siRNA Details
Gene Symbol: Ppa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100834.1 | NP_001094304.1 | 4 | 339 | ||
GenBank mRNA | BC099794.1 | - | 4 | 339 | ||
FQ215196.1 | - | 4 | 291 | |||
FQ215675.1 | - | 4 | 291 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
