siRNA Details
Gene Symbol: Pald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034128.2 | NP_001029300.2 | 10 | 1304 | ||
XM_006256446.2 | XP_006256508.1 | 11 | 2079 | |||
XM_006256447.2 | XP_006256509.1 | 11 | 1515 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
