siRNA Details
Gene Symbol: Sesn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106396.1 | NP_001099866.1 | 6, 7 | 1559 | ||
XM_006256560.2 | XP_006256622.1 | 8, 9 | 2297 | |||
XM_006256561.2 | XP_006256623.1 | 8, 9 | 1309 |
